Assay Details
| Gene Symbol: LOC100056321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646873.1 | XP_023502641.1 | 530 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100056321| Gene Symbol: LOC100056321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023646873.1 | XP_023502641.1 | 530 | 117 | ||
Set Membership: |
|