Assay Details
| Gene Symbol: LOC100062210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602712.3 | XP_005602769.2 | 4029 | 91 | ||
| XM_023647955.1 | XP_023503723.1 | 4026 | 91 | |||
| XM_023647957.1 | XP_023503725.1 | 3966 | 91 | |||
| XM_023647962.1 | XP_023503730.1 | 3873 | 91 | |||
| XM_023647966.1 | XP_023503734.1 | 3810 | 91 | |||
| GenBank mRNA | JL624164.1 | - | 2 - 3 | 633 | 91 | |
More Information
Set Membership: |
|

LOC100062210
Genomic Map