Assay Details
| Gene Symbol: LOC100062210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005602712.3 | XP_005602769.2 | 884 | 101 | ||
| XM_023647955.1 | XP_023503723.1 | 881 | 101 | |||
| XM_023647957.1 | XP_023503725.1 | 884 | 101 | |||
| XM_023647962.1 | XP_023503730.1 | 884 | 101 | |||
| XM_023647966.1 | XP_023503734.1 | 884 | 101 | |||
More Information
Set Membership: |
|

LOC100062210
Genomic Map