Assay Details
| Gene Symbol: MSMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734115.1 | XP_014589601.1 | 107 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734115.1 transcripts.
Set Membership: |
|

MSMB
Genomic Map