Assay Details
| Gene Symbol: LOC100052045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500694.6 | XP_001500744.1 | 181 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001500694.6 transcripts.
Set Membership: |
|

LOC100052045
Genomic Map