Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023648619.1 | XP_023504387.1 | 2717 | 79 | ||
XM_023648622.1 | XP_023504390.1 | 2658 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
