Assay Details
| Gene Symbol: LOC100065612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499085.6 | XP_001499135.1 | 996 | 87 | ||
| XM_014739894.2 | XP_014595380.1 | 999 | 87 | |||
| XM_023641313.1 | XP_023497081.1 | 997 | 87 | |||
| XM_023641314.1 | XP_023497082.1 | 1000 | 87 | |||
| XM_023641315.1 | XP_023497083.1 | 660 | 87 | |||
More Information
Set Membership: |
|

LOC100065612
Genomic Map