Assay Details
| Gene Symbol: LOC100050004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001487971.4 | XP_001488021.1 | 655 | 72 | ||
| XM_005602865.3 | XP_005602922.1 | 426 | 72 | |||
| XM_023652387.1 | XP_023508155.1 | 1368 | 72 | |||
More Information
Set Membership: |
|

LOC100050004
Genomic Map