Assay Details
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652840.1 | XP_023508608.1 | 862 | 78 | ||
| XM_023652849.1 | XP_023508617.1 | 440 | 78 | |||
| XM_023652857.1 | XP_023508625.1 | 924 | 78 | |||
| XM_023652864.1 | XP_023508632.1 | 927 | 78 | |||
| XM_023652868.1 | XP_023508636.1 | 935 | 78 | |||
| XM_023652869.1 | XP_023508637.1 | 865 | 78 | |||
| XM_023652880.1 | XP_023508648.1 | 289 | 78 | |||
| XM_023652889.1 | XP_023508657.1 | 703 | 78 | |||
| XM_023652896.1 | XP_023508664.1 | 253 | 78 | |||
| XM_023652904.1 | XP_023508672.1 | 292 | 78 | |||
| XM_023652907.1 | XP_023508675.1 | 1922 | 78 | |||
| XM_023652913.1 | XP_023508681.1 | 224 | 78 | |||
| XM_023652919.1 | XP_023508687.1 | 232 | 78 | |||
| XM_023652922.1 | XP_023508690.1 | 256 | 78 | |||
| XM_023652924.1 | XP_023508692.1 | 218 | 78 | |||
| XM_023652925.1 | XP_023508693.1 | 1925 | 78 | |||
| XM_023652929.1 | XP_023508697.1 | 194 | 78 | |||
| XM_023652936.1 | XP_023508704.1 | 100 | 78 | |||
| XM_023652942.1 | XP_023508710.1 | 852 | 78 | |||
| XM_023652952.1 | XP_023508720.1 | 229 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023652840.1, XM_023652849.1, XM_023652857.1, XM_023652864.1, XM_023652868.1, XM_023652869.1, XM_023652880.1, XM_023652889.1, XM_023652896.1, XM_023652904.1, XM_023652907.1, XM_023652913.1, XM_023652919.1, XM_023652922.1, XM_023652924.1, XM_023652925.1, XM_023652929.1, XM_023652936.1, XM_023652942.1, XM_023652952.1 transcripts.
Set Membership: |
|

NRG4
Genomic Map