Assay Details
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652942.1 | XP_023508710.1 | 921 | 76 | ||
| XM_023652952.1 | XP_023508720.1 | 298 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023652942.1, XM_023652952.1 transcripts.
Set Membership: |
|

NRG4
Genomic Map