Assay Details
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023652840.1 | XP_023508608.1 | 1295 | 96 | ||
| XM_023652849.1 | XP_023508617.1 | 873 | 96 | |||
| XM_023652857.1 | XP_023508625.1 | 1357 | 96 | |||
| XM_023652864.1 | XP_023508632.1 | 1360 | 96 | |||
| XM_023652868.1 | XP_023508636.1 | 1368 | 96 | |||
| XM_023652869.1 | XP_023508637.1 | 1298 | 96 | |||
| XM_023652880.1 | XP_023508648.1 | 722 | 96 | |||
| XM_023652889.1 | XP_023508657.1 | 1136 | 96 | |||
| XM_023652896.1 | XP_023508664.1 | 686 | 96 | |||
| XM_023652904.1 | XP_023508672.1 | 725 | 96 | |||
| XM_023652907.1 | XP_023508675.1 | 2355 | 96 | |||
| XM_023652913.1 | XP_023508681.1 | 657 | 96 | |||
| XM_023652919.1 | XP_023508687.1 | 665 | 96 | |||
| XM_023652922.1 | XP_023508690.1 | 689 | 96 | |||
| XM_023652924.1 | XP_023508692.1 | 651 | 96 | |||
| XM_023652925.1 | XP_023508693.1 | 2358 | 96 | |||
| XM_023652929.1 | XP_023508697.1 | 627 | 96 | |||
| XM_023652936.1 | XP_023508704.1 | 533 | 96 | |||
| XM_023652942.1 | XP_023508710.1 | 1172 | 96 | |||
| XM_023652952.1 | XP_023508720.1 | 549 | 96 | |||
More Information
Set Membership: |
|

NRG4
Genomic Map