Assay Details
| Gene Symbol: LOC100069641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014733650.2 | XP_014589136.2 | 1005 | 80 | ||
| XM_023617368.1 | XP_023473136.1 | 824 | 80 | |||
More Information
Set Membership: |
|

LOC100069641
Genomic Map