Assay Details
| Gene Symbol: LOC100056283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503012.5 | XP_001503062.1 | 905 | 70 | ||
| XM_023618755.1 | XP_023474523.1 | 808 | 70 | |||
More Information
Set Membership: |
|

LOC100056283
Genomic Map