Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503635.5 | XP_001503685.1 | 519 | 73 | ||
| XM_014734065.2 | XP_014589551.1 | 526 | 73 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDIN1| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503635.5 | XP_001503685.1 | 519 | 73 | ||
| XM_014734065.2 | XP_014589551.1 | 526 | 73 | |||