Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503635.5 | XP_001503685.1 | 730 | 75 | ||
| XM_014734065.2 | XP_014589551.1 | 737 | 75 | |||
| XM_023620455.1 | XP_023476223.1 | 329 | 75 | |||

CDIN1
Genomic Map