Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503635.5 | XP_001503685.1 | 860 | 91 | ||
| XM_014734065.2 | XP_014589551.1 | 867 | 91 | |||
| XM_023620455.1 | XP_023476223.1 | 459 | 91 | |||
More Information
Set Membership: |
|

CDIN1
Genomic Map