Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001505152.4 | XP_001505202.1 | 243 | 68 | ||
| XM_023621175.1 | XP_023476943.1 | 400 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPPP2| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001505152.4 | XP_001505202.1 | 243 | 68 | ||
| XM_023621175.1 | XP_023476943.1 | 400 | 68 | |||
Set Membership: |
|