Assay Details
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603211.3 | XP_005603268.1 | 1217 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
