Assay Details
| Gene Symbol: BCL2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493694.5 | XP_001493744.1 | 3342 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001493694.5 transcripts.
Set Membership: |
|

BCL2L2
Genomic Map