Assay Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492030.4 | XP_001492080.1 | 815 | 86 | ||
| XM_023622553.1 | XP_023478321.1 | 575 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THTPA| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492030.4 | XP_001492080.1 | 815 | 86 | ||
| XM_023622553.1 | XP_023478321.1 | 575 | 86 | |||
Set Membership: |
|