Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624691.1 | XP_023480459.1 | 601 | 89 | ||
| XM_023624696.1 | XP_023480464.1 | 597 | 89 | |||
| XM_023624704.1 | XP_023480472.1 | 384 | 89 | |||
| XM_023624706.1 | XP_023480474.1 | 380 | 89 | |||
More Information
Set Membership: |
|

LRR1
Genomic Map