Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625231.1 | XP_023480999.1 | 553 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTRAF| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625231.1 | XP_023480999.1 | 553 | 59 | ||
Set Membership: |
|