Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596128.3 | XP_005596185.1 | 2604 | 79 | ||
| XM_005596130.3 | XP_005596187.1 | 4660 | 79 | |||
| XM_014733607.2 | XP_014589093.1 | 4199 | 79 | |||
| XM_014733613.2 | XP_014589099.1 | 4660 | 79 | |||
| XM_014733638.2 | XP_014589124.1 | 4660 | 79 | |||
| XM_023649503.1 | XP_023505271.1 | 1162 | 79 | |||
| XM_023649504.1 | XP_023505272.1 | 1147 | 79 | |||
| XM_023649505.1 | XP_023505273.1 | 709 | 79 | |||

GARRE1
Genomic Map