Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014733276.2 | XP_014588762.1 | 750 | 70 | ||
XM_014733284.2 | XP_014588770.1 | 750 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014733276.2, XM_014733284.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
