Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493662.6 | XP_001493712.3 | 831 | 107 | ||
XM_005596212.3 | XP_005596269.1 | 833 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
