Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493784.5 | XP_001493834.2 | 1070 | 135 | ||
XM_014733148.2 | XP_014588634.1 | 1076 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
