Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651329.1 | XP_023507097.1 | 829 | 93 | ||
| XM_023651330.1 | XP_023507098.1 | 732 | 93 | |||
| XM_023651331.1 | XP_023507099.1 | 741 | 93 | |||
| XM_023651332.1 | XP_023507100.1 | 662 | 93 | |||
More Information
Set Membership: |
|

ZNF570
Genomic Map