Assay Details
| Gene Symbol: ZNF780A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596238.3 | XP_005596295.1 | 167 | 108 | ||
| XM_014735583.2 | XP_014591069.1 | 225 | 108 | |||
| XM_014735591.2 | XP_014591077.1 | 331 | 108 | |||
| XM_023649737.1 | XP_023505505.1 | 372 | 108 | |||
| XM_023649741.1 | XP_023505509.1 | 224 | 108 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596238.3, XM_014735583.2, XM_014735591.2, XM_023649737.1, XM_023649741.1 transcripts.
Set Membership: |
|

ZNF780A
Genomic Map