Assay Details
| Gene Symbol: ZNF780A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596238.3 | XP_005596295.1 | 219 | 81 | ||
| XM_014735583.2 | XP_014591069.1 | 277 | 81 | |||
| XM_014735591.2 | XP_014591077.1 | 383 | 81 | |||
| XM_014735658.2 | XP_014591144.1 | 475 | 81 | |||
| XM_023649737.1 | XP_023505505.1 | 424 | 81 | |||
| XM_023649738.1 | XP_023505506.1 | 622 | 81 | |||
| XM_023649739.1 | XP_023505507.1 | 581 | 81 | |||
| XM_023649740.1 | XP_023505508.1 | 417 | 81 | |||
| XM_023649741.1 | XP_023505509.1 | 276 | 81 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596238.3, XM_014735583.2, XM_014735591.2, XM_014735658.2, XM_023649737.1, XM_023649738.1, XM_023649739.1, XM_023649740.1, XM_023649741.1 transcripts.
Set Membership: |
|

ZNF780A
Genomic Map