Assay Details
| Gene Symbol: ZNF780A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596238.3 | XP_005596295.1 | 349 | 101 | ||
| XM_005596242.3 | XP_005596299.1 | 382 | 101 | |||
| XM_014735583.2 | XP_014591069.1 | 407 | 101 | |||
| XM_014735591.2 | XP_014591077.1 | 513 | 101 | |||
| XM_014735658.2 | XP_014591144.1 | 605 | 101 | |||
| XM_023649737.1 | XP_023505505.1 | 554 | 101 | |||
| XM_023649738.1 | XP_023505506.1 | 752 | 101 | |||
| XM_023649739.1 | XP_023505507.1 | 711 | 101 | |||
| XM_023649740.1 | XP_023505508.1 | 547 | 101 | |||
| XM_023649741.1 | XP_023505509.1 | 406 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596242.3 transcripts.
Set Membership: |
|

ZNF780A
Genomic Map