Assay Details
| Gene Symbol: LOC100069923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JL620847.1 | - | 1 - 2 | 463 | 81 | |
| KT124644.1 | - | 2 - 3 | 438 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100069923| Gene Symbol: LOC100069923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JL620847.1 | - | 1 - 2 | 463 | 81 | |
| KT124644.1 | - | 2 - 3 | 438 | 81 | ||
Set Membership: |
|