Assay Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023649860.1 | XP_023505628.1 | 664 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
