Assay Details
| Gene Symbol: GPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729383.2 | XP_014584869.2 | 363 | 70 | ||
| XM_023649920.1 | XP_023505688.1 | 980 | 70 | |||
| XM_023649921.1 | XP_023505689.1 | 177 | 70 | |||
| XM_023649922.1 | XP_023505690.1 | 296 | 70 | |||
| XM_023649923.1 | XP_023505691.1 | 246 | 70 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014729383.2, XM_023649920.1, XM_023649921.1, XM_023649922.1, XM_023649923.1 transcripts.
Set Membership: |
|

GPR4
Genomic Map