Assay Details
| Gene Symbol: ODAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650018.1 | XP_023505786.1 | 1666 | 92 | ||
| XM_023650019.1 | XP_023505787.1 | 1543 | 92 | |||
| XM_023650020.1 | XP_023505788.1 | 1663 | 92 | |||
| XM_023650021.1 | XP_023505789.1 | 1660 | 92 | |||
More Information
Set Membership: |
|

ODAD1
Genomic Map