Assay Details
| Gene Symbol: LOC100066474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650305.1 | XP_023506073.1 | 835 | 85 | ||
| XM_023650306.1 | XP_023506074.1 | 793 | 85 | |||
| XM_023650307.1 | XP_023506075.1 | 502 | 85 | |||
More Information
Set Membership: |
|

LOC100066474
Genomic Map