Assay Details
| Gene Symbol: LOC100054321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596524.3 | XP_005596581.1 | 1377 | 59 | ||
| XM_023650415.1 | XP_023506183.1 | 1377 | 59 | |||
More Information
Set Membership: |
|

LOC100054321
Genomic Map