Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650571.1 | XP_023506339.1 | 780 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023650571.1 transcripts.
Set Membership: |
|

CCDC106
Genomic Map