Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650712.1 | XP_023506480.1 | 6338 | 137 | ||
| XM_023650713.1 | XP_023506481.1 | 6386 | 137 | |||
| XM_023650715.1 | XP_023506483.1 | 6231 | 137 | |||
| XM_023650716.1 | XP_023506484.1 | 6311 | 137 | |||
| XM_023650717.1 | XP_023506485.1 | 6308 | 137 | |||
| XM_023650718.1 | XP_023506486.1 | 6305 | 137 | |||
| XM_023650719.1 | XP_023506487.1 | 6338 | 137 | |||
| XM_023650720.1 | XP_023506488.1 | 6099 | 137 | |||
| XM_023650721.1 | XP_023506489.1 | 6275 | 137 | |||
| XM_023650722.1 | XP_023506490.1 | 6126 | 137 | |||
| XM_023650723.1 | XP_023506491.1 | 6239 | 137 | |||
| XM_023650724.1 | XP_023506492.1 | 6099 | 137 | |||
More Information
Set Membership: |
|

DOP1A
Genomic Map