Assay Details
| Gene Symbol: LOC102147786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596868.3 | XP_005596925.3 | 3540 | 91 | ||
| XM_023650884.1 | XP_023506652.1 | 4809 | 91 | |||
| XM_023650885.1 | XP_023506653.1 | 4695 | 91 | |||
| XM_023650886.1 | XP_023506654.1 | 4677 | 91 | |||
More Information
Set Membership: |
|

LOC102147786
Genomic Map