Assay Details
| Gene Symbol: DDO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001916542.4 | XP_001916577.2 | 1239 | 72 | ||
| XM_014734560.2 | XP_014590046.1 | 654 | 72 | |||
| XM_014734561.2 | XP_014590047.1 | 605 | 72 | |||
| XM_014734564.2 | XP_014590050.1 | 654 | 72 | |||
| XM_014734567.2 | XP_014590053.1 | 610 | 72 | |||
More Information
Set Membership: |
|

DDO
Genomic Map