Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502358.6 | XP_001502408.3 | 528 | 60 | ||
| XM_014734653.2 | XP_014590139.1 | 797 | 60 | |||
| XM_023650953.1 | XP_023506721.1 | 455 | 60 | |||
| XM_023650954.1 | XP_023506722.1 | 800 | 60 | |||
More Information
Set Membership: |
|

CCN6
Genomic Map