Assay Details
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502358.6 | XP_001502408.3 | 970 | 83 | ||
| XM_023650953.1 | XP_023506721.1 | 897 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502358.6 | XP_001502408.3 | 970 | 83 | ||
| XM_023650953.1 | XP_023506721.1 | 897 | 83 | |||
Set Membership: |
|