Assay Details
Gene Symbol: C5H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610330.3 | XP_005610387.1 | 250 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005610330.3 transcripts.
Set Membership: |
![]() ![]() |
