Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490335.5 | XP_001490385.5 | 1017 | 121 | ||
| XM_023641564.1 | XP_023497332.1 | 1017 | 121 | |||
More Information
Set Membership: |
|

PLPPR5
Genomic Map