Assay Details
| Gene Symbol: RFPL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502418.3 | XP_001502468.1 | 248 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502418.3 transcripts.
Set Membership: |
|

RFPL4B
Genomic Map