Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490394.6 | XP_001490444.3 | 700 | 80 | ||
| XM_014735780.2 | XP_014591266.1 | 700 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490394.6 | XP_001490444.3 | 700 | 80 | ||
| XM_014735780.2 | XP_014591266.1 | 700 | 80 | |||