Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597106.3 | XP_005597163.1 | 616 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597106.3 transcripts.
Set Membership: |
|

SEPTIN9
Genomic Map