Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597105.2 | XP_005597162.1 | 1334 | 107 | ||
| XM_005597106.3 | XP_005597163.1 | 1921 | 107 | |||
| XM_005597107.3 | XP_005597164.1 | 1453 | 107 | |||
| XM_005597110.3 | XP_005597167.1 | 799 | 107 | |||
| XM_023651940.1 | XP_023507708.1 | 1952 | 107 | |||
| XM_023651941.1 | XP_023507709.1 | 1636 | 107 | |||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map