Assay Details
| Gene Symbol: LOC100060051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736574.2 | XP_014592060.2 | 812 | 92 | ||
| XM_023652051.1 | XP_023507819.1 | 790 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100060051| Gene Symbol: LOC100060051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736574.2 | XP_014592060.2 | 812 | 92 | ||
| XM_023652051.1 | XP_023507819.1 | 790 | 92 | |||