Assay Details
| Gene Symbol: C11H17orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597224.3 | XP_005597281.1 | 169 | 115 | ||
| XM_005597226.3 | XP_005597283.1 | 168 | 115 | |||
| XM_005597227.3 | XP_005597284.1 | 168 | 115 | |||
| XM_023652118.1 | XP_023507886.1 | 170 | 115 | |||
| XM_023652119.1 | XP_023507887.1 | 169 | 115 | |||
| XM_023652121.1 | XP_023507889.1 | 169 | 115 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597224.3, XM_005597226.3, XM_005597227.3, XM_023652118.1, XM_023652119.1, XM_023652121.1 transcripts.
Set Membership: |
|

C11H17orf80
Genomic Map